missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD1B Antibody (OTI1C7), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73945AF700
This item is not returnable.
View return policy
Description
RPRD1B Monoclonal specifically detects RPRD1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RPRD1B | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RPRD1B | |
| Full length recombinant protein of human RPRD1B (NP_067038) produced in HEK293T cells. | |
| 0.1 mL | |
| 58490.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1C7 | |
| Western Blot, Immunohistochemistry | |
| Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction