missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD1B Antibody (OTI1C7) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73945
This item is not returnable.
View return policy
Description
RPRD1B Monoclonal specifically detects RPRD1B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RPRD1B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:4000, Immunohistochemistry 1:150 | |
| Cell cycle-related and expression-elevated protein in tumor, cell-cycle related and expression-elevated protein in tumor, chromosome 20 open reading frame 77, CREPTC20orf77, dJ1057B20.2, DKFZp434P0735, FLJ44520, NET60, regulation of nuclear pre-mRNA domain containing 1B, regulation of nuclear pre-mRNA domain-containing protein 1B | |
| Mouse | |
| 36.7 kDa | |
| 100 μg | |
| 58490 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot, Immunohistochemistry | |
| OTI1C7 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| RPRD1B | |
| Full length human recombinant protein of human RPRD1B(NP_067038) produced in HEK293T cell. | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction