missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPRD2 Antibody [Janelia Fluor™ 585], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-97932JF585
This item is not returnable.
View return policy
Description
RPRD2 Polyclonal antibody specifically detects RPRD2 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| RPRD2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Janelia Fluor 585 | |
| HSPC099, KIAA0460FLJ32145, regulation of nuclear pre-mRNA domain containing 2, regulation of nuclear pre-mRNA domain-containing protein 2 | |
| The immunogen this antibody was made to, maps to a region between residues 1000 and 1050 of human RPRD2 using the numbering given in TrEMBL entry Q5VT52 (GeneID 23248). | |
| 0.1 mL | |
| DNA Repair | |
| 23248 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction