missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS15A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561426

Product Code. 15759241

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: TGRLNKCGVI SPRFDVQLKD LE Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

RPS15A consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8P family of ribosomal proteins. It is located in the cytoplasm. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RPS15A
Polyclonal
Unconjugated
RPS15A
40S ribosomal protein S15a; A630031B11Rik; OK/SW-cl.82; ribosomal protein S15a; Rps15a; S15a; Small ribosomal subunit protein uS8; up-regulated by HBV X protein
Rabbit
Antigen affinity chromatography
RUO
6210
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P62244
RPS15A
Recombinant protein corresponding to Human RPS15A. Recombinant protein control fragment (Product #RP-100802).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.