missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS19 Polyclonal Antibody

RPS19 Polyclonal Antibody
Brand: Invitrogen PA575564
This item is not returnable.
View return policy
Description
RPS19 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| RPS19 | |
| Polyclonal | |
| Unconjugated | |
| RPS19 | |
| 40S ribosomal protein S19; DBA; DBA1; Dsk3; HGNC:10402; ribosomal protein S19; Rps19; S19; Small ribosomal subunit protein eS19 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 108348115, 20085, 29287, 6223 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17074, P39019, Q9CZX8 | |
| RPS19 | |
| Synthetic peptide corresponding to amino acids 100-145 of Human RPS19. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction