missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599706
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Ribosomal Protein S2.
Bacterial pathogens deliver type III effector proteins into the plant cell during infection. Plants express disease resistance (R) proteins that respond specifically to a particular type III effector by activating immune responses. Resistance proteins guard the plant against pathogens that contain an appropriate avirulence protein via an indirect interaction. RPS2 (resistance to Pseudomonas syringae protein 2) is a 909 amino acid plasma membrane protein with leucine-rich repeat, leucine zipper and P loop domains that confers resistance to Pseudomonas syringae infection by interacting with the avirulence gene AvrRpt2. Belonging to the disease resistance NB-LRR family, RPS2 directly associates with RIN4, which triggers plant resistance when RIN4 is degraded by AvrRpt2.
Specifications
| RPS2 | |
| Polyclonal | |
| Unconjugated | |
| Rps2 | |
| 40S ribosomal protein S2; 40S ribosomal protein S4; fc94f01; hypothetical protein LOC406565; ik:tdsubc_2h2; Llrep3; OK/KNS-cl.6; protein LLRep3; repeat family 3; ribosomal protein S2; ribosomal protein S2e; rps2; rps2e; Rps2r1; RPS4; S2; S4; Small ribosomal subunit protein uS5; tdsubc_2h2; wu:fb10e01; wu:fc94f01; wu:fj09b04; xx:tdsubc_2h2; zgc:85824 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6187, 83789 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P15880, P27952 | |
| Rps2 | |
| A synthesized peptide derived from human RPS2(Accession P15880), corresponding to amino acid residues K257-T293. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction