missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS24 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93809-0.02ml
This item is not returnable.
View return policy
Description
RPS24 Polyclonal antibody specifically detects RPS24 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| RPS24 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunoprecipitation 1:50 - 1:100, Immunohistochemistry-Paraffin | |
| DBA3,40S ribosomal protein S24, DKFZp686N1586, ribosomal protein S24 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human RPS24 (NP_148982.1). MNDTVTIRTRKFMTNRLLQRKQMVIDVLHPGKATVPKTEIREKLAKMYKTTPDVIFVFGFRTHFGGGKTTGFGMIYDSLDYAKKNEPKHRLARHGLYEKKKTSRKQRKERKNRMKKVRGTAKANVGAGKK | |
| 0.02 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6229 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction