missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RPS24 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15799860 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15799860 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15799860 Supplier Invitrogen™ Supplier No. PA551911

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TNRLLQRKQM VIDVLHPGKA TVPKTEIREK LAKMYKTTPD VIFVFGFRTH FGGGKTTGFG MIYDSLDYAK KNEPKHRLAR HGLYEKKKTS RKQRKERKNR MK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS24 is a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S24E family of ribosomal proteins. It is located in the cytoplasm. Multiple transcript variants encoding different isoforms have been found for this gene. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RPS24
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RPS24
Gene Accession No. P62847
Gene Alias 40S ribosomal protein S24; DBA3; DKFZp686N1586; MRP S24; ribosomal protein S24; ribosomal protein S24, isoform 1; RPS24; Rps24 protein; S24; Small ribosomal subunit protein eS24; wu:fa66b02; zgc:103596; zgc:109740
Gene Symbols RPS24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RPS24. Recombinant protein control fragment (Product #RP-102208).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6229
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.