missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS27L Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94193-0.1ml
This item is not returnable.
View return policy
Description
RPS27L Polyclonal antibody specifically detects RPS27L in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RPS27L | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 40S ribosomal protein S27-like, ribosomal protein S27-like | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-84 of human RPS27L (NP_057004.1). MPLARDLLHPSLEEEKKKHKKKRLVQSPNSYFMDVKCPGCYKITTVFSHAQTVVLCVGCSTVLCQPTGGKARLTEGCSFRRKQH | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Direct Reversal of DNA Damage | |
| 51065 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction