missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS27L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570649
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 100%.
Ubiquitin, a highly conserved protein that has a major role in targeting cellular proteins for degradation by the 26S proteosome, is synthesized as a precursor protein consisting of either polyubiquitin chains or a single ubiquitin fused to an unrelated protein. This gene encodes a fusion protein consisting of ubiquitin at the N terminus and ribosomal protein S27a at the C terminus. When expressed in yeast, the protein is post-translationally processed, generating free ubiquitin monomer and ribosomal protein S27a. Ribosomal protein S27a is a component of the 40S subunit of the ribosome and belongs to the S27AE family of ribosomal proteins. It contains C4-type zinc finger domains and is located in the cytoplasm. Pseudogenes derived from this gene are present in the genome. As with ribosomal protein S27a, ribosomal protein L40 is also synthesized as a fusion protein with ubiquitin; similarly, ribosomal protein S30 is synthesized as a fusion protein with the ubiquitin-like protein fubi. Multiple alternatively spliced transcript variants that encode the same proteins have been identified.
Specifications
| RPS27L | |
| Polyclonal | |
| Unconjugated | |
| RPS27L | |
| 1810034D23Rik; 40S ribosomal protein S27-like; 40S ribosomal protein S27-like protein; OTTHUMP00000200786; OTTHUMP00000201692; ribosomal protein S27; ribosomal protein S27 like; ribosomal protein S27-like; rps27l; Small ribosomal subunit protein eS27-like | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 681429 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P24051 | |
| RPS27L | |
| synthetic peptide directed towards the middle region of rat RPS27L. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction