missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ RPS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30704
This item is not returnable.
View return policy
Description
RPS3 Polyclonal antibody specifically detects RPS3 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| RPS3 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| 40S ribosomal protein S3, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, ribosomal protein S3 | |
| Between 1 and 50 | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6188 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction