missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS3 Antibody (RP159-1), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80043UV
This item is not returnable.
View return policy
Description
RPS3 Monoclonal antibody specifically detects RPS3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RPS3 | |
| Monoclonal | |
| DyLight 350 | |
| 40S ribosomal protein S3, FLJ26283, FLJ27450, IMR-90 ribosomal protein S3, MGC87870, ribosomal protein S3 | |
| Recombinant human RPS3. | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 6188 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| RP159-1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction