missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS4X Antibody [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-21449JF669
This item is not returnable.
View return policy
Description
RPS4X Polyclonal antibody specifically detects RPS4X in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RPS4X | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Janelia Fluor 669 | |
| CCG2Single copy abundant mRNA protein, DXS306, FLJ40595, ribosomal protein S4, X-linked, RPS4cell cycle gene 2, SCAR40S ribosomal protein S4, X isoform, SCR10ribosomal protein S4X isoform, single-copy abundant mRNA | |
| Produced in rabbits immunized with E. coli-derived Human RPS4X fragment | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 6191 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction