missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS5 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97627G
This item is not returnable.
View return policy
Description
RPS5 Polyclonal specifically detects RPS5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RPS5 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| ribosomal protein S5,40S ribosomal protein S5 | |
| Produced in rabbits immunized with E. coli-derived Human RPS5 fragment. | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 6193 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction