missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6, Rabbit anti-Human, Clone: CD10, Abnova™
Rabbit Monoclonal Antibody
Brand: Abnova MAB19062.20uL
This item is not returnable.
View return policy
Specifications
| RPS6 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA). | |
| - | |
| Rabbit | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| CD10 | |
| Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user. | |
| ribosomal protein S6 | |
| RPS6 | |
| A synthetic phosphopeptide corresponding to residues surrounding S240/S244 of human RPS6. | |
| RUO | |
| 6194 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction