missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KA2 Monoclonal Antibody (OTI5G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809874S
This item is not returnable.
View return policy
Description
RPS6KA2 Monoclonal antibody specifically detects RPS6KA2 in Human samples. It is validated for Western BlotSpecifications
| RPS6KA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q15349 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6196 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5G2 | |
| Unconjugated | |
| RPS6KA2 | |
| 90 kDa ribosomal protein S6 kinase 2; 90kDa; D17Wsu134e; HU-2; MAP kinase-activated protein kinase 1c; MAPK-activated protein kinase 1c; MAPKAP kinase 1C; MAPKAPK1C; MAPKAPK-1c; mitogen-activated protein kinase-activated protein kinase 1C; p90rsk; p90-RSK 2; p90RSK2; p90-RSK3; pp90rsk; pp90RSK3; Protein-tyrosine kinase Mpk-9; ribosomal protein S6 kinase A2; ribosomal protein S6 kinase alpha 2; ribosomal protein S6 kinase alpha-2; ribosomal protein S6 kinase polypeptide 2; ribosomal protein S6 kinase, 90kD, polypeptide 2; ribosomal protein S6 kinase, 90kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; ribosomal S6 kinase 3; Rps6ka2; Rps6ka-rs1; RSK; Rsk3; RSK-3; S6K-alpha; S6K-alpha 2; S6K-alpha2; S6K-alpha-2 | |
| Human recombinant protein fragment corresponding to amino acids 1-51 of human RPS6KA2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction