missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RPS6KB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582389
This item is not returnable.
View return policy
Description
Immunogen sequence: VDSPDDTALS ESANQAFLGF TYVAPSVLDS IKEGFSFQPK LRSPRRLNSS PRAPVSPLKF SPFEGFRPSP SLPEPTELPL PPLLPPPPPS TTAPLPIRPP SGTKK Highest antigen sequence indentity to the following orthologs: Rat - 89%, Mouse - 88%.
RPS6KB2 (p70S6K beta) is a serine/threonine kinase with 2 non-identical kinase catalytic domains which phosphorylates the S6 ribosomal protein and eukaryotic translation initiation factor 4B (eIF4B). Phosphorylation of S6 leads to an increase in protein synthesis and cell proliferation.
Specifications
| RPS6KB2 | |
| Polyclonal | |
| Unconjugated | |
| Rps6kb2 | |
| 70 kDa ribosomal protein S6 kinase 2; 70kDa; KLS; p70 ribosomal S6 kinase beta; p70 S6 kinase beta; p70 S6KB; p70 S6K-beta; p70(S6K)-beta; P70-beta; P70-beta-1; P70-beta-2; p70-S6K 2; P70S6K2; p70S6Kb; ribosomal protein S6 kinase B2; ribosomal protein S6 kinase beta-2; ribosomal protein S6 kinase, 70kD, polypeptide 2; ribosomal protein S6 kinase, 70kDa, polypeptide 2; ribosomal protein S6 kinase, polypeptide 2; RPS6KB2; S6 kinase 2; S6 kinase-related kinase; S6K2; S6K-beta; S6K-beta2; S6K-beta-2; serine/threonine-protein kinase 14 beta; serine/threonine-protein kinase 14B; SRK; STK14B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 361696, 58988, 6199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UBS0, Q9Z1M4 | |
| Rps6kb2 | |
| Recombinant protein corresponding to Human RPS6KB2. Recombinant protein control fragment (Product #RP-88709). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction