missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS6KC1 Antibody (OTI10F11), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73946FR
This item is not returnable.
View return policy
Description
RPS6KC1 Monoclonal specifically detects RPS6KC1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| RPS6KC1 | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| RPS6KC1 | |
| Recombinant protein fragment corresponding to amino acids 1-305 of human RPS6KC1 (NP_036556) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI10F11 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11.1, humS6PKh1, ribosomal protein S6 kinase delta-1, ribosomal protein S6 kinase, 52kD, polypeptide 1,52 kDa ribosomal protein S6 kinase, ribosomal protein S6 kinase, 52kDa, polypeptide 1, Ribosomal S6 kinase-like protein with two PSK domains 118 kDa protein, RPK118, S6K-delta-1, SPHK1-binding protein | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 26750.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction