missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS8, Mouse anti-Human, Clone: 4D11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006202-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: YRQWYESHYALPLGRKKGAKLTPEEEEILNKKRSKKIQKKYDERKKNAKISSLLEEQFQQGKLLACIASRPGQCGRADGYVLEGKELEFYLRKIKARKGSpecifications
| RPS8 | |
| Monoclonal | |
| Unconjugated | |
| ribosomal protein S8 | |
| - | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4D11 | |
| In 1x PBS, pH 7.4 | |
| NM_001012 | |
| RPS8 | |
| RPS8 (NP_001003.1, 109 a.a. ∼ 207 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6202 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction