Learn More
Invitrogen™ RPS8 Recombinant Rabbit Monoclonal Antibody (24GB4120)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557228
 View more versions of this product
Product Code. 30276518
  • € 151.00 / 20µL
Description
RPS8 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S8E family of ribosomal proteins. It is located in the cytoplasm. Increased expression of this gene in colorectal tumors and colon polyps compared to matched normal colonic mucosa has been observed. This gene is co-transcribed with the small nucleolar RNA genes U38A, U38B, U39, and U40, which are located in its fourth, fifth, first, and second introns, respectively. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RPS8
Recombinant Monoclonal
Unconjugated
Rps8
40S ribosomal protein S8; OK/SW-cl.83; ribosomal protein S8; Rps8; S8; Small ribosomal subunit protein eS8
Rabbit
Affinity chromatography
RUO
20116, 6202, 65136
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4120
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P62241, P62242, P62243
Rps8
A synthesized peptide derived from human RPS8 (aa 1-208).