missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS9 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22298APC
This item is not returnable.
View return policy
Description
RPS9 Polyclonal antibody specifically detects RPS9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation (Negative)
Specifications
| RPS9 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoprecipitation (Negative) | |
| ribosomal protein S9,40S ribosomal protein S9 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ribosomal Protein S9 using the numbering given in entry NP_001004.2 (GeneID 6203). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| APC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6203 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction