missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPSA Antibody (MLuC5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-631
This item is not returnable.
View return policy
Description
RPSA Monoclonal specifically detects RPSA in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RPSA | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1:10 - 1:200, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:200, Immunohistochemistry-Paraffin 4 - 8 ug/mL | |
| 37 kDa laminin receptor precursor, 37/67 kDa laminin receptor, 37LRPNEM/1CHD4, 67 kDa laminin receptor, 67kD, ribosomal protein SA, 67LR, Colon carcinoma laminin-binding protein, LAMBR37 kDa laminin receptor, laminin binding protein, Laminin receptor 1, laminin receptor 1 (67kD, ribosomal protein SA), Laminin-binding protein precursor p40, lamR, LAMR 1 LAMR140S ribosomal protein SA, LBP, LBP/p40, LRP, LRP/LR, Multidrug resistance-associated protein MGr1-Ag, p40, ribosomal protein SA | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Precipitation |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| MLuC5 | |
| Unconjugated | |
| P08865 | |
| RPSA | |
| Small cell lung carcinoma N592 live cells | |
| 500 μL | |
| Core ESC Like Genes, Cytoskeleton Markers, Extracellular Matrix, Neuroscience, Stem Cell Markers | |
| 3921 | |
| Store at 4C. Do not freeze. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction