missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPTOR Antibody (514208) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5957X
This item is not returnable.
View return policy
Description
RPTOR Monoclonal antibody specifically detects RPTOR in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| RPTOR | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 514208 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
| E. coli-derived recombinant human RPTOR, Lys77-Gln230, Accession # Q8N122 | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
| 57521 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction