missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPTOR Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-766G
This item is not returnable.
View return policy
Description
RPTOR Polyclonal antibody specifically detects RPTOR in Human samples. It is validated for Western Blot
Specifications
| RPTOR | |
| Polyclonal | |
| Western Blot | |
| KIAA1303p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, KOG1, Mip1, p150 target of rapamycin (TOR)-scaffold protein, Raptor, regulatory associated protein of MTOR, complex 1, regulatory-associated protein of mTOR | |
| The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human Regulatory Associated Protein of mTOR using the numbering given in entry AAM09075.1 (GeneID 57521). | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway, Signal Transduction | |
| 57521 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction