missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RQ-00203078, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
RQ-00203078 is a highly selective, potent and orally active TRPM8 antagonist with IC50s of 5.3 nM and 8.3 nM for rat and human TRPM8 channels, respectively. RQ-00203078 shows little inhibitory action against TRPV1, TRPA1, TRPV4, or TRPM2 channels.
Specifications
Specifications
| Chemical Name or Material | RQ-00203078 |
| CAS | 1254205-52-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H13ClF6N2O5S |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 38 mg/mL (68.49 mM) |
| SMILES | O=C(O)C1=CC=C(S(=O)(N(C2=NC=C(C(F)(F)F)C=C2Cl)CC3=CC=C(OC(F)(F)F)C=C3)=O)C=C1 |
| Molecular Weight (g/mol) | 554.85 |
| Formula Weight | 554.85 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction