missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RRAGA Polyclonal Antibody
GREENER_CHOICE

Product Code. 16619414
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16619414 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16619414 Supplier Invitrogen™ Supplier No. PA5102957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RRAGA.

RRAGA (Ras Related GTP Binding A) is a Protein Coding gene. Diseases associated with RRAGA includeFamilial Renal OncocytomaandTuberous Sclerosis 1. Among its related pathways aremTOR signallingandMetabolism of proteins. Gene Ontology (GO) annotations related to this gene includeprotein homodimerization activityandGTP binding. An important paralog of this gene isRRAGB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RRAGA
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene RRAGA
Gene Accession No. Q63486, Q7L523, Q80X95
Gene Alias 1300010C19Rik; Adenovirus E3 14.7 kDa-interacting protein 1; adenovirus E3-14.7K interacting protein 1; AI255374; FIP1; FIP-1; Rag A; RAGA; Ras related GTP binding A; Ras-related GTP binding A; ras-related GTP-binding protein A; Ras-related GTP-binding protein ragA; RGD:619804}; rraga; rraga {ECO:0000312
Gene Symbols RRAGA
Host Species Rabbit
Immunogen A synthesized peptide derived from human RRAGA(Accession Q7L523), corresponding to amino acid residues S277-R313.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10670, 117044, 68441
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.