missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAGC Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32202B
This item is not returnable.
View return policy
Description
RRAGC Polyclonal antibody specifically detects RRAGC in Human, Mouse samples. It is validated for Western Blot
Specifications
| RRAGC | |
| Polyclonal | |
| Western Blot | |
| FLJ13311, GTPase-interacting protein 2, GTR2, Rag C, Rag C protein, RAGC, Ras-related GTP binding C, ras-related GTP-binding protein C, TIB929 | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Ras-Related GTP Binding C using the numbering given in entry NP_071440.1 (GeneID 64121). | |
| 0.1 mL | |
| Apoptosis, mTOR Pathway | |
| 64121 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction