missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRAS Antibody (S06-7A2), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | RRAS |
|---|---|
| Clone | S06-7A2 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot 1:1000 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684963
|
Novus Biologicals
NBP3-19764-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18657503
|
Novus Biologicals
NBP3-19764-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RRAS Monoclonal antibody specifically detects RRAS in Human samples. It is validated for Western BlotSpecifications
| RRAS | |
| 0.3 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 6237 | |
| IgG | |
| Affinity purified |
| S06-7A2 | |
| Western Blot 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Oncogene RRAS, p23, ras-related protein R-Ras, related RAS viral (r-ras) oncogene homolog | |
| A synthetic peptide of human RRAS (Uniprot # P10301) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title