missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RRAS2 Monoclonal Antibody (EM-50)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528616
This item is not returnable.
View return policy
Description
This antibody recognizes R-Ras2 / TC21 protein (intracellular antigen) and does not react with R-Ras1, H-Ras, K-Ras, and N-Ras.
RRAS2 is involved in activating mutations and the overexpression of classical Ras subfamily members (K-RAS, N-RAS and H-RAS) have been widely investigated as key events in the development of human cancers. The RRAS subfamily of Ras-related proteins includes RRAS1, RRAS2 (TC21) and RRAS3 (M-Ras) show overall amino acid identity with the classical Ras subfamily (H-Ras, K-Ras and N-Ras) of 55-60%. RRAS2 is a small GTP binding protein of the Ras superfamily of GTPases. It might transduce growth inhibitory signals across the cell membrane, exerting its effect through an effector shared with the Ras proteins. RRAS2 has high oncogenic potential and overexpression/mutations have been reported in several tumor tissues and cell lines.
Specifications
| RRAS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P62070 | |
| RRAS2 | |
| Human TC21. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
| EM-50 | |
| Unconjugated | |
| RRAS2 | |
| 2610016H24Rik; C86394; Ras-like protein TC21; ras-related protein R-Ras2; related RAS viral (r-ras) oncogene homolog 2; Rras2; TC21; Teratocarcinoma oncogene | |
| Mouse | |
| Protein A | |
| RUO | |
| 22800 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction