missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM1 Antibody (OTI5G5), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73947DL594
This item is not returnable.
View return policy
Description
RRM1 Monoclonal antibody specifically detects RRM1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| RRM1 | |
| Monoclonal | |
| DyLight 594 | |
| EC 1.17.4.1, R1, ribonucleoside-diphosphate reductase large subunit, Ribonucleoside-diphosphate reductase subunit M1, Ribonucleotide reductase large subunit, ribonucleotide reductase M1, ribonucleotide reductase M1 polypeptide, ribonucleotide reductase, large subunit, ribonucleotide reductase, R1 subunit, RIR1, RR1 | |
| Recombinant protein fragment corresponding to amino acids 541-792 of human RRM1 (NP_001024) produced in E.coli. | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 6240 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5G5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction