missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM1 Monoclonal Antibody (OTI12B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804483
This item is not returnable.
View return policy
Description
RRM1 (Ribonucleotide reductase M1 polypeptide) is one of two non-identical subunits that constitute ribonucleoside-diphosphate reductase, an enzyme which catalyzes the biosynthesis of deoxyribonucleotides from the corresponding ribonucleotides. It provides the precursors necessary for DNA synthesis. RRM1 is involved in carcinogenesis, tumor progression.Specifications
| RRM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23921 | |
| RRM1 | |
| Human recombit protein fragment corresponding to amino acids 541-792 of human RRM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI12B9 | |
| Unconjugated | |
| RRM1 | |
| R1, RIR1, RR1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6240 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction