missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRM1 Monoclonal Antibody (OTI12B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA804483S
This item is not returnable.
View return policy
Description
RRM1 Monoclonal antibody specifically detects RRM1 in Human samples. It is validated for Western BlotSpecifications
| RRM1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06702 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 6280 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI12B9 | |
| Unconjugated | |
| S100A9 | |
| cb396; cb838; CHUNP6866; hypothetical protein; R RRM 1; r1; RI RI RR 1; Ribonucleoside-diphosphate reductase large subunit; ribonucleoside-diphosphate reductase M1 chain; ribonucleoside-diphosphate reductase subunit M1; ribonucleotide reductase catalytic subunit M1; ribonucleotide reductase large subunit; ribonucleotide reductase M1; ribonucleotide reductase M1 L homeolog; ribonucleotide reductase M1 polypeptide; ribonucleotide reductase M1 S homeolog; ribonucleotide reductase protein R1 class I; ribonucleotide reductase subunit M1; ribonucleotide reductase, R1 subunit; RIR 1; RIR1; RnrM1; RR1; Rrm1; rrm1 protein; rrm1.L; rrm1.S; sb:cb548; wu:fb39b07; wu:fi14b02; wu:fk95f07; XELAEV_18013932mg; XRRM1 | |
| Full length human recombinant protein of human S100A9 produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction