missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85339-25ul
This item is not returnable.
View return policy
Description
RRP1 Polyclonal specifically detects RRP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| RRP1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| D21S2056ENop52, NNP1, NNP-1Nnp1 homolog, nucleolar protein, NOP52protein NNP-1, Novel nuclear protein 1, Nucleolar protein Nop52, ribosomal RNA processing 1 homolog (S. cerevisiae), RRP1Aribosomal RNA processing protein 1 homolog A, RRP1-like protein | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8568 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| RRP1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GSICRAEPEAGEEQAGDDRDSGGPVLQFDYEAVANRLFEMASRQSTPSQNRKRLYKVIRKLQDLAGGIFPEDEIPEKACRRLLEGRRQKK | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction