missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP12 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36547B
This item is not returnable.
View return policy
Description
RRP12 Polyclonal antibody specifically detects RRP12 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RRP12 | |
| Polyclonal | |
| Western Blot | |
| FLJ20231, KIAA0690DKFZp762P1116, ribosomal RNA processing 12 homolog (S. cerevisiae), RRP12-like protein | |
| The immunogen recognized by the antibody maps to a region between residue 1247 to 1297 of human Ribosomal RNA Processing 12 Homolog using the numbering given in entry NP_055994.2. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23223 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction