Learn More
RRP12 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-36547C
Product Code. 18770141
  • € 633.00 / 0.10mL
Description
RRP12 Polyclonal antibody specifically detects RRP12 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RRP12
DyLight 650
The immunogen recognized by the antibody maps to a region between residue 1247 to 1297 of human Ribosomal RNA Processing 12 Homolog using the numbering given in entry NP_055994.2.
Polyclonal
Rabbit
Store at 4°C in the dark.