Learn More
RRP12 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-36547C
This item is not returnable.
View return policy
Description
RRP12 Polyclonal antibody specifically detects RRP12 in Human, Mouse samples. It is validated for Western Blot
Specifications
| RRP12 | |
| DyLight 650 | |
| The immunogen recognized by the antibody maps to a region between residue 1247 to 1297 of human Ribosomal RNA Processing 12 Homolog using the numbering given in entry NP_055994.2. |
| Polyclonal | |
| Rabbit | |
| Store at 4°C in the dark. |