missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 470.00
Specifications
| Antigen | RRP9 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626502
|
Novus Biologicals
NBP2-94262-0.02ml |
0.02 mL |
€ 190.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661722
|
Novus Biologicals
NBP2-94262-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RRP9 Polyclonal antibody specifically detects RRP9 in Mouse samples. It is validated for Western BlotSpecifications
| RRP9 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| ribosomal RNA processing 9, small subunit (SSU) processome component, homolog(yeast), RNA, U3 small nucleolar interacting protein 2, RNU3IP2, RRP9 homolog, U3 small nucleolar ribonucleoprotein-associated 55 kDa protein, U3 small nucleolar RNA-interacting protein 2, U3 snoRNP-associated 55 kDa protein, U3 snoRNP-associated 55-kDa protein, U355K, U3-55KRRP9, small subunit (SSU) processome component, homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 156-475 of human RRP9 (NP_004695.1). PDDSAIFSAAKDCSIIKWSVESGRKLHVIPRAKKGAEGKPPGHSSHVLCMAISSDGKYLASGDRSKLILIWEAQSCQHLYTFTGHRDAVSGLAFRRGTHQLYSTSHDRSVKVWNVAENSYVETLFGHQDAVAALDALSRECCVTAGGRDGTVRVWKIPEESQLVFYGHQGSIDCIHLINEEHMVSGADDGSVALWGLSKKRPLALQREAHGLRGEPGLEQPFWISSVAALLNTDLVATGSHSSCVRLWQCGEGFRQLDLLCDIPLVGFINSLKFSSSGDFLVAGVGQEHRLGRWWRIKEARNSVCIIPLRRVPVPPAAGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9136 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts