missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-4-[4-[(Oxiran-2-yl)methoxy]-1,2,5-thiadiazol-3-yl]morpholine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
(RS)-4-[4-[(Oxiran-2-yl)methoxy]-1,2,5-thiadiazol-3-yl]morpholine
Specifications
Specifications
| Chemical Name or Material | rac 4-[4-(Oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]morpholine |
| CAS | 58827-68-2 |
| Analyte or Component Names | rac 4-[4-(Oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]morpholine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H13 N3 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Morpholine, 4-[4-(2-oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]-, Morpholine, 4-[4-(oxiranylmethoxy)-1,2,5-thiadiazol-3-yl]- (9CI), 1,2,5-Thiadiazole, morpholine deriv., 4-[4-[(Oxiran-2-yl)methoxy]-1,2,5-thiadiazol-3-yl]morpholine |
| InChI Formula | InChI=1S/C9H13N3O3S/c1-3-13-4-2-12(1)8-9(11-16-10-8)15-6-7-5-14-7/h7H,1-6H2 |
| SMILES | C(Oc1nsnc1N2CCOCC2)C3CO3 |
| IUPAC Name | 4-[4-(oxiran-2-ylmethoxy)-1,2,5-thiadiazol-3-yl]morpholine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction