missing translation for 'onlineSavingsMsg'
Learn More

(RS)-N-(3-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride, Mikromol™

Product Code. 30052208
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052208 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30052208 Supplier Mikromol Supplier No. MM0563.04

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(RS)-N-(3-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (RS)-N-(3-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride
CAS 878791-35-6
Analyte or Component Names (RS)-N-(3-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O . Cl H
Certifications/Compliance ISO 17025
Synonym Prilocaine Hydrochloride Imp. D (EP), Prilocaine Imp. D (EP), (RS)-N-(3-Methyl-phenyl)-2-(propylamino)propanamide Hydrochloride, Prilocaine Hydrochloride Imp. D (EP) as Hydrochloride, Prilocaine Hydrochloride Impurity D as Hydrochloride, Prilocaine Impurity D as Hydrochloride
InChI Formula InChI=1S/C13H20N2O.ClH/c1-4-8-14-11(3)13(16)15-12-7-5-6-10(2)9-12;/h5-7,9,11,14H,4,8H2,1-3H3,(H,15,16);1H
SMILES Cl.CCCNC(C)C(=O)Nc1cccc(C)c1
IUPAC Name N-(3-methylphenyl)-2-(propylamino)propanamide;hydrochloride
Molecular Weight (g/mol) 256.77
Quantity 50 mg
Formula Weight 256.1342 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.