Learn More
RSAD2 Mouse, anti-Human, Clone: 1C5D4, Proteintech
Product Code. 18020626 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18020626 20 μL 20µL
18085285 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18020626 Supplier Proteintech Supplier No. 686041IG20UL
Mouse Monoclonal Antibody
The RSAD2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RSAD2. This antibody recognizes human, mouse, pig antigen. The RSAD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Antigen RSAD2
Applications Western Blot, ELISA
Clone 1C5D4
Concentration 1000 μg/mL
Gene Symbols 2510004L01Rik, cig33, cig5, RSAD2, vig1, Viperin
Host Species Mouse
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 91543
Target Species Human, Mouse, Pig
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG1