missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSBN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559879
This item is not returnable.
View return policy
Description
Immunogen sequence: KEILTQGQIN STSGLNKESF RYLKDEQLCR LNLGMQEYRV PQGVQTPFMT HQEHSIRRNF LKTGTKFSNF IHEEHQSNGG ALV Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Histone demethylase that specifically demethylates dimethylated 'Lys-20' of histone H4 (H4K20me2), thereby modulating chromosome architecture. [UniProt].
Specifications
| RSBN1 | |
| Polyclonal | |
| Unconjugated | |
| RSBN1 | |
| C230004D03Rik; E330004N01; KDM9; Kiaa3002; Lysine-specific demethylase 9; mKIAA3002; RGD1307231; Rosbin; rosbin, round spermatid basic protein 1; round spermatid basic protein 1; Rsbn; Rsbn1; Rsbp | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54665 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5VWQ0 | |
| RSBN1 | |
| Recombinant protein corresponding to Human RSBN1. Recombinant protein control fragment (Product #RP-99854). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction