Learn More
RSK1, p Ser380 Antibody (1024F), Alexa Fluor™ 594, R&D Systems™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals MAB79672AF594
This item is not returnable.
View return policy
Description
RSK1 Monoclonal specifically detects RSK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RSK1 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| RPS6KA1 | |
| Phosphopeptide containing the human RSK1 S380 site, Accession No. Q15418 | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1024F | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunocytochemistry | |
| 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1 | |
| Rabbit | |
| Protein A or G purified | |
| MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
| 6195.0 | |
| Store at 4°C in the dark. |