missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK1, p Thr359, p Ser363 Antibody (S01-5B1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20023-50ul
This item is not returnable.
View return policy
Description
RSK1 Monoclonal antibody specifically detects RSK1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| RSK1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-5B1 | |
| Unconjugated | |
| 90kD, polypeptide 1), EC 2.7.11, EC 2.7.11.1,90 kDa ribosomal protein S6 kinase 1, HU-1, MAP kinase-activated protein kinase 1a, MAPK-activated protein kinase 1a, MAPKAP kinase 1a, MAPKAPK-1a, p90RSK1, p90S6K, ribosomal protein S6 kinase alpha 1, ribosomal protein S6 kinase alpha-1, ribosomal protein S6 kinase, 90kD, polypeptide 1, ribosomal protein S6 kinase, 90kDa, polypeptide 1, Ribosomal S6 kinase 1, RSK, RSK-1, RSK1p90-RSK 1, S6K-alpha 1, S6K-alpha-1 | |
| A synthetic phosphopeptide corresponding to residues surrounding Thr359/Ser363 of human RSK1 (Uniprot # Q15418) | |
| 50 μg | |
| MAP Kinase Signaling, mTOR Pathway, Phospho Specific, Protein Kinase, Signal Transduction | |
| 6195 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction