missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSK1 Recombinant Rabbit Monoclonal Antibody (JE48-76)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA534863
This item is not returnable.
View return policy
Description
Positive Control: Daudi cell, Hela cell, rat testis tissue, human appendix tissue, human prostate cancer tissue, human small intestine tissue, mouse colon tissue, F9 cell.
RPS6KA1 (RSK1) is a growth-factor regulated serine/threonine kinase that is involved in the MAPK cascade. It is thought to control cellular proliferation and differentiation through the phosphorylation of transcription factors. RPS6KA1 contains 2 nonidentical kinase catalytic domains and phosphorylates various substrates, including members of the mitogen-activated kinase (MAPK) signalling pathway. The activity of this protein has been implicated in controlling cell growth and differentiation. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| RSK1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q15418 | |
| RPS6KA1 | |
| Recombinant protein within Human RSK1 p90 aa 1-107. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| JE48-76 | |
| Unconjugated | |
| RPS6KA1 | |
| 90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinase, polypeptide A1; ribosomal S6 kinase 1; RPS6KA; Rps6ka1; rps6ka1.L; RPS6KA1L; rsk; RSK1; Rsk-1; S6 kinase II beta; S6 protein kinase (Rsk-1); S6K-alpha 1; S6K-alpha-1; S6KII-alpha; S6KII-beta; wu:fc56h07; XELAEV_18012109mg; zgc:152654 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6195 | |
| -20°C, Avoid Freeze/Thaw Cycles, store in dark | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction