missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48825
This item is not returnable.
View return policy
Description
RSK3 Polyclonal antibody specifically detects RSK3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RSK3 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| EC 2.7.11, EC 2.7.11.1, HU-2, MAP kinase-activated protein kinase 1c, MAPK-activated protein kinase 1c, MAPKAP kinase 1c, MAPKAPK-1c, MAPKAPK1C, p90-RSK 2, p90RSK2, p90-RSK3, ribosomal protein S6 kinase alpha 2, ribosomal protein S6 kinase alpha-2, ribosomal protein S6 kinase, 90kD, polypeptide 2, ribosomal protein S6 kinase, 90kDa, polypeptide 2, Ribosomal S6 kinase 3,90 kDa ribosomal protein S6 kinase 2, RSK, RSK-3, RSK3pp90RSK3, S6K-alpha, S6K-alpha2, S6K-alpha-2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: MDLSMKKFAVRRFFSVYLRRKSRSKSSSLSRLEEEGVVKEIDISHHVKEGF | |
| 0.1 mL | |
| Protein Kinase | |
| 6196 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction