missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSL1D1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97626MFV500
This item is not returnable.
View return policy
Description
RSL1D1 Polyclonal antibody specifically detects RSL1D1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| RSL1D1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| CATX11, CATX-11, Cellular senescence-inhibited gene protein, CSIGMGC142259, DKFZP564M182, L12, MGC138433, PBK1cellular senescence inhibited gene protein, Protein PBK1, ribosomal L1 domain containing 1, ribosomal L1 domain-containing protein 1 | |
| Produced in rabbits immunized with E. coli-derived Human RSL1D1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 26156 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction