missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: KERKKKRQQA RKTASVLSKD DVAPESGDTT VKKPESKKEQ TPEHGKKKRG RGKAQVKATN ESEDEIPQLV PIGKKTPANE KVEIQKHATG KKS Highest antigen sequence identity to the following orthologs: Mouse - 35%, Rat - 38%.
Regulates cellular senescence through inhibition of PTEN translation. Acts as a pro-apoptotic regulator in response to DNA damage.
Specifications
Specifications
| Antigen | RSL1D1 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RSL1D1 |
| Gene Accession No. | O76021 |
| Gene Alias | 2410005K20Rik; Ac1158; C76439; CATX11; CATX-11; cellular senescence inhibited; cellular senescence inhibited gene protein; cellular senescence-inhibited gene protein; CSIG; DKFZP564M182; L12; MGC138433; MGC142259; pBK1; Protein PBK1; ribosomal L1 domain containing 1; ribosomal L1 domain-containing protein 1; RSL1D1; UTP30 |
| Gene Symbols | RSL1D1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
