missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSL24D1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564058
This item is not returnable.
View return policy
Description
Immunogen sequence: QRELWNKTID AMKRVEEIKQ KRQAKFIMNR LKKNKELQKV QDIKEVKQNI H Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 98%.
This gene encodes a protein sharing a low level of sequence similarity with human ribosomal protein L24. Although this gene has been referred to as RPL24, L30, and 60S ribosomal protein L30 isolog in the sequence databases, it is distinct from the human genes officially named RPL24 (which itself has been referred to as ribosomal protein L30) and RPL30. The protein encoded by this gene localizes to the nucleolus and is thought to play a role in the biogenesis of the 60S ribosomal subunit. The precise function of this gene is currently unknown. This gene utilizes alternative polyadenylation signals and has multiple pseudogenes.
Specifications
| RSL24D1 | |
| Polyclonal | |
| Unconjugated | |
| RSL24D1 | |
| 2410159K22Rik; 60S ribosomal protein L30 isolog; C15orf15; homolog of yeast ribosomal like protein 24; HRP-L30-iso; L30; My024; my024 protein; probable ribosome biogenesis protein RLP24; ribosomal L24 domain containing 1; ribosomal L24 domain-containing protein 1; Ribosomal protein L24-like; RLP24; RPL24; RPL24L; RSL24D1; TVAS3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51187 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UHA3 | |
| RSL24D1 | |
| Recombinant protein corresponding to Human RSL24D1. Recombinant protein control fragment (Product #RP-103466). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction