missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RSL24D1 Polyclonal Antibody

Description
Immunogen sequence: SGPIYPGHGM MFVRNDCKVF RFCKSKCHKN FKKKRNPRKV RWTKAFRKAA GKELTVDNSF EFEKRRNEPI K.
This gene encodes a protein sharing a low level of sequence similarity with human ribosomal protein L24. Although this gene has been referred to as RPL24, L30, and 60S ribosomal protein L30 isolog in the sequence databases, it is distinct from the human genes officially named RPL24 (which itself has been referred to as ribosomal protein L30) and RPL30. The protein encoded by this gene localizes to the nucleolus and is thought to play a role in the biogenesis of the 60S ribosomal subunit. The precise function of this gene is currently unknown. This gene utilizes alternative polyadenylation signals and has multiple pseudogenes.
Specifications
Specifications
| Antigen | RSL24D1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 2.83 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | RSL24D1 |
| Gene Accession No. | Q99L28, Q9UHA3 |
| Gene Alias | 2410159K22Rik; 60S ribosomal protein L30 isolog; C15orf15; homolog of yeast ribosomal like protein 24; HRP-L30-iso; L30; My024; my024 protein; probable ribosome biogenesis protein RLP24; ribosomal L24 domain containing 1; ribosomal L24 domain-containing protein 1; Ribosomal protein L24-like; RLP24; RPL24; RPL24L; RSL24D1; TVAS3 |
| Gene Symbols | RSL24D1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?