missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSL24D1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25190-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| RSL24D1 | |
| Polyclonal | |
| Unconjugated | |
| RSL24D1 | |
| C15orf15, HRP L30 iso, L30, Ribosomal protein L24 like, RLP24, RPL24, RPL24L, RSL24D1, TVAS3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51187 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9UHA3 | |
| RSL24D1 | |
| RSL24D1 Fusion Protein Ag18195 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction