Learn More
RSPO3 Mouse anti-Human, CoraLite 488, Clone: 1E4F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-66314-100UL
Product Code. 16890766
  • € 450.00 / 100µL
Description
The RSPO3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RSPO3. This antibody recognizes human antigen. The RSPO3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
Specifications
RSPO3
Monoclonal
CoraLite 488
100 μL
84870
IgG2b
Immunofluorescence
1E4F1